EMAIL THIS PAGE TO A FRIEND
Clinical science (London, England : 1979)
Increased galectin-3 levels are associated with abdominal aortic aneurysm progression and inhibition of galectin-3 decreases elastase-induced AAA development.
PMID 28982723
Abstract
Abdominal aortic aneurysm (AAA) evolution is unpredictable and no specific treatment exists for AAA, except surgery to prevent aortic rupture. Galectin-3 has been previously associated with CVD, but its potential role in AAA has not been addressed. Galectin-3 levels were increased in the plasma of AAA patients (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1250 Elastase from porcine pancreas, Type I, ≥4.0 units/mg protein