EMAIL THIS PAGE TO A FRIEND
International journal of cardiology
Myeloid-related protein-8/14 in acute coronary syndrome.
PMID 28986057
Abstract
The alarmin family member myeloid-related protein (MRP)-14 (S100A9), which has been identified by platelet transcriptional profiling as an acute myocardial infarction gene, regulates vascular inflammation and thrombosis. Elevated plasma levels of MRP-8/14 (S100A8/A9) heterodimer predict first and recurrent cardiovascular events. The aim of this study was to elucidate pathophysiological roles of MRP-8/14 in acute coronary syndrome (ACS). In 38 consecutive ACS patients, the MRP-8/14 level in coronary artery blood obtained at thrombus aspiration was higher in 23 patients, in whom aspirated thrombus was confirmed, compared to the 15 patients, in whom it was absent [4.86 (1.95, 8.29) vs 2.94 (1.31, 4.44), P=0.017]. The MRP-8/14 level was correlated with myeloperoxidase (MPO) level (R Our findings indicate that MRP-8/14 concentration increases in coronary artery blood in association with thrombus formation in ACS, co-localizes with leukocytes, and is associated with leukocyte activation. MRP-8/14 is positioned as a unique biomarker at the interface of inflammation and thrombosis in ACS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0642 Human F3 / Tissue Factor ELISA Kit