EMAIL THIS PAGE TO A FRIEND
Cell
Structural Basis for a Safety-Belt Mechanism That Anchors Condensin to Chromosomes.
PMID 28988770
Abstract
Condensin protein complexes coordinate the formation of mitotic chromosomes and thereby ensure the successful segregation of replicated genomes. Insights into how condensin complexes bind to chromosomes and alter their topology are essential for understanding the molecular principles behind the large-scale chromatin rearrangements that take place during cell divisions. Here, we identify a direct DNA-binding site in the eukaryotic condensin complex, which is formed by its Ycg1
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34025
Dibromobimane, BioReagent, suitable for fluorescence, ≥95.0% (CHN)
C10H10Br2N2O2