EMAIL THIS PAGE TO A FRIEND
Chinese journal of natural medicines
Protosappanin A exerts anti-neuroinflammatory effect by inhibiting JAK2-STAT3 pathway in lipopolysaccharide-induced BV2 microglia.
PMID 28991528
Abstract
Microglial activation and resultant neuroinflammatory response are implicated in various brain diseases including Alzheimer's disease and Parkinson's disease. Treatment with anti-neuroinflammatory agents could provide therapeutic benefits for such disorders. Protosappanin A (PTA) is a major bioactive ingredient isolated from Caesalpinia sappan L.. In this work, the anti-neuroinflammatory effects of PTA on LPS-stimulated BV2 cells were investigated and the underlying mechanisms were explored. Results showed that PTA significantly inhibited the production of TNF-α and IL-1β in LPS-activated BV2 microglia. Moreover, the mRNA expressions of IL-6, IL-1β, and MCP-1 were reduced by PTA in a dose-dependent manner. Furthermore, PTA suppressed JAK2/STAT3-dependent inflammation pathway through down-regulating the phosphorylation of JAK2 and STAT3, as well as STAT3 nuclear translocation against LPS treatment. These observations suggested a novel role for PTA in regulating LPS-induced neuroinflammatory injuries.