EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Analysis and Sensory Evaluation of Volatile Constituents of Fresh Blackcurrant (Ribes nigrum L.) Fruits.
PMID 28992408
Abstract
Volatile constituents of fresh blackcurrant (Ribes nigrum L.) berries were isolated via vacuum-headspace extraction and analyzed by capillary gas chromatography-mass spectrometry. In agreement with previous studies with frozen fruits, short-chain esters and terpenes were major compound classes. However, rather high concentrations of C
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31140
L-(+)-Fructose, ≥97.0% (HPLC)
C6H12O6