EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Rhodol-derived Colorimetric and Fluorescent Probe with the Receptor of Carbonothioate for the Specific Detection of Mercury Ions.
PMID 28993592
Abstract
Developing some methods that can simply and effectively detect mercury ions (Hg