EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
The Oleaginous Yeast
PMID 29018411
Abstract
A high throughput screening (HTS) methodology for evaluation of cellular lipid content based on Nile red fluorescence reads using black background 96-wells test plates and a plate reader equipment allowed the rapid intracellular lipid estimation of strains from a Brazilian phylloplane yeast collection. A new oleaginous yeast,