EMAIL THIS PAGE TO A FRIEND
Clinical epigenetics
Down-regulation of MBD4 contributes to hypomethylation and overexpression of CD70 in CD4
PMID 29018507
Abstract
Systemic lupus erythematosus (SLE) is an autoimmune disease that is characterized by lymphocytic infiltration and overproduction of autoantibodies, leading to significant morbidity and mortality. However, the pathogenesis of this disorder has not yet been completely elucidated. It has been reported that CD70, a B cell costimulatory molecule encoded by the gene Our results showed that the expression of MBD4 was significantly decreased in CD4 Our results indicated that downregulation of MBD4 contributed to overexpression and hypomethylation of the CD70 gene in SLE CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU025861 MISSION® esiRNA, esiRNA human MBD4 (esiRNA1)