EMAIL THIS PAGE TO A FRIEND
Molecular cancer therapeutics
PPARγ Ligand-induced Annexin A1 Expression Determines Chemotherapy Response via Deubiquitination of Death Domain Kinase RIP in Triple-negative Breast Cancers.
PMID 29021293
Abstract
Metastatic breast cancer is still incurable so far; new specifically targeted and more effective therapies for triple-negative breast cancer (TNBC) are required in the clinic. In this study, our clinical data have established that basal and claudin-low subtypes of breast cancer (TNBC types) express significantly higher levels of Annexin A1 (ANXA1) with poor survival outcomes. Using human cancer cell lines that model the TNBC subtype, we observed a strong positive correlation between expression of ANXA1 and PPARγ. A similar correlation between these two markers was also established in our clinical breast cancer patients' specimens. To establish a link between these two markers in TNBC, we show