EMAIL THIS PAGE TO A FRIEND
Scientific reports
Development of a Cell-Based Assay for Measuring Base Excision Repair Responses.
PMID 29021553
Abstract
Base excision repair (BER) is the predominant pathway for coping with most forms of hydrolytic, oxidative or alkylative DNA damage. Measuring BER capacity in living cells is valuable for both basic science applications and epidemiological studies, since deficiencies in this pathway have been associated with cancer susceptibility and other adverse health outcomes. At present, there is an ongoing effort to develop methods to effectively quantify the rate of BER as a whole. We present a variation of a previously described "Oligonucleotide Retrieval Assay" designed to measure DNA excision repair that is capable of quantifying the rate of repair of thymine glycol in a variety of human cells with a high degree of sensitivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU154471 MISSION® esiRNA, esiRNA human NTHL1 (esiRNA1)