EMAIL THIS PAGE TO A FRIEND
Cell death & disease
Differential regulation of spermatogenic process by Lkb1 isoforms in mouse testis.
PMID 29022902
Abstract
Liver serine/threonine kinase B1 (LKB1) is a tumor suppressor associated with the pathogenesis of Peutz-Jeghers syndrome. Affected males are at increased risk of developing Sertoli cell tumors and display defective spermatogenesis. Male mice lacking the short isoform (Lkb1
Related Materials