EMAIL THIS PAGE TO A FRIEND
Chemosphere
Electrospun polycaprolactam-manganese oxide fiber for headspace-solid phase microextraction of phthalate esters in water samples.
PMID 29028539
Abstract
The nanofibrous polycaprolactam (polyamide 6 (PA6)) incorporated with manganese oxide (MnO) nanoparticles was fabricated by electrospinning and used as a new fiber coating for headspace-solid phase microextraction (HS-SPME) of the selected phthalate esters (PEs) in water samples prior to GC-μECD. The adsorbent was fully characterized using scanning electron microscopy (SEM), Fourier transform-infrared (FT-IR) spectroscopy and thermogravimetric analysis (TGA). The main parameters that affect the HS-SPME efficiency such as extraction temperature, ionic strength, extraction and desorption times were investigated. The analytical figures of merit were obtained under the optimized conditions as follows: linear dynamic range (LDR), 0.500-5.00 × 10