EMAIL THIS PAGE TO A FRIEND
Oncotarget
RNAi targeting Nogo Receptor enhanced survival and proliferation of murine retinal ganglion cells during N-methyl-D-aspartate-induced optic nerve crush.
PMID 29029408
Abstract
We investigated the effects of lentivirus-mediated RNAi targeting of Nogo Receptor (