EMAIL THIS PAGE TO A FRIEND
Oncotarget
The immune impact of mimic endoscopic retrograde appendicitis therapy and appendectomy on rabbits of acute appendicitis.
PMID 29029533
Abstract
This study was conducted to evaluate the immune impact of mimic endoscopic retrograde appendicitis therapy and appendectomy on rabbits of acute suppurative appendicitis and to determine whether TLR4/MYD88/NF-κB signaling pathway was activated in this process. 48 rabbits were assigned into 4 groups: group I, the mimic endoscopic retrograde appendicitis therapy group; group II, the appendectomy group; group III, the model group; and group IV, the blank group. White blood cells decreased, while levels of C-reactive protein, tumor necrosis factor-α, interleukin-6, interleukin-4, and interleukin-10 increased on the 2
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0797 Mouse IgA ELISA Kit