EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Thermal desorption comprehensive two-dimensional gas chromatography coupled to time of flight mass spectrometry for vapour phase mainstream tobacco smoke analysis.
PMID 29030031
Abstract
A thermal desorption comprehensive two-dimensional gas chromatography coupled to time-of-flight mass spectrometry (TD-GC×GC-TOFMS) method has been developed for the analysis of mainstream tobacco smoke (MTS) vapour phase (VP). The selection process of the sample introduction approach involved comparing the results obtained from three different approaches: a) use of gas sampling bag followed by SPME (Tedlar
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25461 Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
24241-U Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
25467 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
24246 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
54041-U Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
29753-U Carboxen® 569, O.D. × L 1/4  in. × 3 1/2 , glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29534-U Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
54293-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: (300 mg), cartridge volume 3 mL, pkg of 10 ea
54294-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
10478-U Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
11048-U Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
10477-U Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
11324-U Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
11072-U Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
23446-U ORBO-93 Sorbent Tube, matrix Carboxen® 1000, ORBO tube I.D. × L 6 mm × 95 mm, Bed B 90 mg, Bed A 180 mg, 60/80 mesh, pkg of 25 ea
20355 ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
20358 ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
28718-U Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4  in. × 3 1/2  in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea