EMAIL THIS PAGE TO A FRIEND
The Journal of steroid biochemistry and molecular biology
Applicability of an innovative steroid-profiling method to determine synthetic growth promoter abuse in cattle.
PMID 29030156
Abstract
A robust LC-MS/MS method was developed to quantify a large number of phase I and phase II steroids in urine. The decision limit is for most compounds lower than 1ngml
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
681059
2-Methyl-6-nitrobenzoic anhydride, 97%
C16H12N2O7
R0500
Reichstein’s substance S, ≥98%
C21H30O4