EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Revealing the electronic character of the positive electrode/electrolyte interface in lithium-ion batteries.
PMID 29034947
Abstract
High voltage operating active materials are among the most promising components for positive electrodes of future high energy lithium-ion batteries. However, the operating potential range of such materials often exceeds anodically the thermodynamic stability window of the electrolyte. A surface layer is therefore formed, which is supposed to be one of the reasons for the high irreversible charge loss of these electrodes. The electronic character of such a surface layer formed at the electrode/electrolyte interface of LiNi
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388297
Ferrocenium hexafluorophosphate, 97%
C10H10F6FeP