EMAIL THIS PAGE TO A FRIEND
Food chemistry
Hexafluoroisopropanol-induced salt-free catanionic surfactant coacervate extraction method for determination of fluoroquinolones in milk samples.
PMID 29037667
Abstract
Coacervation and phase separation were first reported in salt-free catanionic surfactant aqueous systems based on lauric acid (LA) and dodecyltrimethylammonium hydroxide (DTAOH), using hexafluoroisopropanol (HFIP) as a coacervate-inducing agent. The liquid-liquid two-phase separation occurs over a wide of LA/DTAOH molar ratios (78:22-0:100mol/mol) and total surfactant concentrations (5-200mmolL
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33700
Danofloxacin, VETRANAL, analytical standard
C19H20FN3O3