EMAIL THIS PAGE TO A FRIEND
Food chemistry
Straightforward analytical method to determine opium alkaloids in poppy seeds and bakery products.
PMID 29037712
Abstract
A straightforward method to determine the content of six opium alkaloids (morphine, codeine, thebaine, noscapine, papaverine and narceine) in poppy seeds and bakery products was developed and validated down to a limit of quantification (LOQ) of 0.1mg/kg. The method was based on extraction with acetonitrile/water/formic acid, ten-fold dilution and analysis by LC-MS/MS using a pH 10 carbonate buffer. The method was applied for the analysis of 41 samples collected in 2015 in the Netherlands and Germany. All samples contained morphine ranging from 0.2 to 240mg/kg. The levels of codeine and thebaine ranged from below LOQ to 348mg/kg and from below LOQ to 106mg/kg, respectively. Sixty percent of the samples exceeded the guidance reference value of 4mg/kg of morphine set by BfR in Germany, whereas 25% of the samples did not comply with the limits set for morphine, codeine, thebaine and noscapine by Hungarian legislation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N9007
Noscapine hydrochloride hydrate, analytical standard
C22H23NO7 · HCl · xH2O