EMAIL THIS PAGE TO A FRIEND
Endocrine connections
GATA transcription factors in testicular adrenal rest tumours.
PMID 29038332
Abstract
Testicular adrenal rest tumours (TARTs) are benign adrenal-like testicular tumours that frequently occur in male patients with congenital adrenal hyperplasia. Recently, GATA transcription factors have been linked to the development of TARTs in mice. The aim of our study was to determine GATA expression in human TARTs and other steroidogenic tissues. We determined GATA expression in TARTs (