EMAIL THIS PAGE TO A FRIEND
Scientific reports
Performance and consistency of a fluorescence-based high-throughput screening assay for use in Babesia drug screening in mice.
PMID 29038534
Abstract
In this study, we evaluated the validity of a fluorescence-based assay using SYBR Green I (SG I) stain for screening antibabesial compounds against B. microti in mice. Two different hematocrits (HCTs; 2.5% and 5%) were used. Correlating relative fluorescence units (RFUs) with parasitemia showed significant linear relationships with R
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7770
Diminazene aceturate, analytical standard
C14H15N7 · 2C4H7NO3
P0049
Pyronaridine tetraphosphate, ≥95% (HPLC)
C29H32ClN5O2·4H3PO4