EMAIL THIS PAGE TO A FRIEND
Nature communications
Direct visualization of critical hydrogen atoms in a pyridoxal 5'-phosphate enzyme.
PMID 29038582
Abstract
Enzymes dependent on pyridoxal 5'-phosphate (PLP, the active form of vitamin B