EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Apurinic/apyrimidinic endonuclease 1 (APE1) contributes to resveratrol‑induced neuroprotection against oxygen‑glucose deprivation and re‑oxygenation injury in HT22 cells: Involvement in reducing oxidative DNA damage.
PMID 29039534
Abstract
Resveratrol, a naturally occurring polyphenolic compound, exhibits a neuroprotective role in models of central nervous system diseases, including cerebral ischemia/reperfusion injury. Apurinic/apyrimidinic endonuclease 1 (APE1) is a multifunctional enzyme that contributes to base excision repair of oxidative DNA damage and redox activation of transcription factors, associated with neuronal survival against hypoxic‑ischemic injury. It was hypothesized that resveratrol protects HT22 cells against oxygen‑glucose deprivation and re‑oxygenation (OGD/R)‑induced injuries through upregulation of APE1. It was demonstrated that resveratrol pretreatment significantly increased the viability of HT22 cells and decreased the release of lactate dehydrogenase (LDH), accompanied by the upregulation of APE1 mRNA, and protein levels, as well as the activity of APE1 under OGD/R conditions. In addition, resveratrol reversed OGD/R‑induced oxidative DNA damage as evidenced by the decreases in the levels of 8‑hydroxy‑2'‑deoxyguanosine and APE sites. However, APE1 knockdown using short hairpin RNA sequence targeting APE1 abolished resveratrol‑elicited beneficent effects against OGD/R‑induced cytotoxicity and oxidative stress. This was indicated by decreased cell viability, superoxide dismutase activity and glutathione levels, and increased LDH release and reactive oxygen species levels. The results of the present study indicate that APE1 contributes to the protective effects of resveratrol against neonatal hypoxic‑ischemic brain injuries, and suggest that DNA repair enzymes, including APE1, may be a unique strategy for neuroprotection against this disease.