EMAIL THIS PAGE TO A FRIEND
Nature communications
Growth mechanism of strongly emitting CH
PMID 29042559
Abstract
Metal halide perovskite nanocrystals are promising materials for a diverse range of applications, such as light-emitting devices and photodetectors. We demonstrate the bandgap tunability of strongly emitting CH
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
806498
Methylammonium bromide
CH6BrN