EMAIL THIS PAGE TO A FRIEND
Infection and drug resistance
Investigating a rare methicillin-resistant
PMID 29042801
Abstract
Methicillin resistant Four CC15-MRSA isolates collected in Riyadh, Saudi Arabia, between 2013 and 2014 were studied. Two isolates were from clinical infection and 2 from retail meat products. Whole genome sequencing was carried out using Illumina HiSeq2500 genome analyzer. All the CC15-MRSA isolates had the multilocus sequence typing profile ST1535, 13-13-1-1-81-11-13, which is a single locus variant of ST15. Of the 6 contigs related to the SCC element, one comprised a recombinase gene CC15-MRSA shows a novel SCC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51387
MRSA Selective Supplement, for microbiology
C17H20N2O6S