EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
On the rate-determining step of fatty acid oxidation in heart. Inhibition of fatty acid oxidation by 4-pentenoic acid.
PMID 29043
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245925
4-Pentenoic acid, 97%
C5H8O2