EMAIL THIS PAGE TO A FRIEND
PeerJ
Mysterious inhibitory cell regulator investigated and found likely to be secretogranin II related.
PMID 29043108
Abstract
In the context of a hunt for a postulated hormone that is tissue-mass inhibiting and reproductively associated, there is described probable relatedness to a granin protein. A 7-8 kDa polypeptide candidate (gels/MS) appeared in a bioassay-guided fractionation campaign involving sheep plasma. An