EMAIL THIS PAGE TO A FRIEND
Journal of experimental botany
Interaction between calcium and potassium modulates elongation rate in cotton fiber cells.
PMID 29045717
Abstract
Calcium (Ca2+) is necessary for fiber cell development in cotton (Gossypium hirsutum), both as a cell wall structural component and for environmental signaling responses. It is also known that potassium (K+) plays a critical role in cotton fiber cell elongation. However, it is unclear whether Ca2+ integrates its activities with K+ to regulate fiber elongation. Here, we report the novel discovery that Ca2+ deficiency, when integrated with K+ signaling, promotes fiber elongation. Using inductively coupled plasma-mass spectrometry (ICP-MS), we determined dynamic profiles of the ionome in ovules and fibers at different developmental stages, and found that a high accumulation of macro-elements, but not Ca2+, was associated with longer fibers. Using an in vitro ovule culture system, we found that under Ca2+-deficient conditions, sufficient K+ (52 mM) rapidly induced ovule and fiber browning, while reduced K+ (2 or 27 mM) not only suppressed tissue browning but also altered fiber elongation. Reduced K+ also enhanced reactive oxygen species scavenging ability and maintained abscisic acid and jasmonic acid levels, which in turn compensated for Ca2+ deficiency. Ca2+ deficiency combined with reduced K+ (0 mM Ca2+ and 27 mM K+) produced longer fibers in cultured ovules, due to cell wall loosening by phytosulfokine (PSK), expansin (EXP), and xyloglucan endotransglycosylase/hydrolase (XTH), and an increase of the K+ content of fiber cells. Using transgenic cotton, we showed that the CBL-INTERACTING PROTEIN KINASE 6 (GhCIPK6) gene mediates the uptake of K+ under Ca2+-deficient conditions. This study establishes a new link between Ca2+, K+, and fiber elongation.