EMAIL THIS PAGE TO A FRIEND
Frontiers in microbiology
Boeravinone B, A Novel Dual Inhibitor of NorA Bacterial Efflux Pump of
PMID 29046665
Abstract
This study elucidated the role of boeravinone B, a NorA multidrug efflux pump inhibitor, in biofilm inhibition. The effects of boeravinone B plus ciprofloxacin, a NorA substrate, were evaluated in NorA-overexpressing, wild-type, and knocked-out
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
96844
Boeravinone B, analytical standard
C17H12O6