EMAIL THIS PAGE TO A FRIEND
Frontiers in immunology
Defining Genome-Wide Expression and Phenotypic Contextual Cues in Macrophages Generated by Granulocyte/Macrophage Colony-Stimulating Factor, Macrophage Colony-Stimulating Factor, and Heat-Killed Mycobacteria.
PMID 29046677
Abstract
Heat-killed (HK)