EMAIL THIS PAGE TO A FRIEND
Nanoscale
Solution-processed indium oxide electron transporting layers for high-performance and photo-stable perovskite and organic solar cells.
PMID 29048085
Abstract
Interface engineering is considered the key to improving the device performance and stability of solar cells. In particular, TiO