EMAIL THIS PAGE TO A FRIEND
Oncotarget
Von Hippel-Lindau regulates interleukin-32β stability in ovarian cancer cells.
PMID 29050245
Abstract
Hypoxia-induced interleukin-32β (IL-32β) shifts the metabolic program to the enhanced glycolytic pathway. In the present study, the underlying mechanism by which hypoxia-induced IL-32β stability is regulated was investigated in ovarian cancer cells. IL-32β expression increased under hypoxic conditions in ovarian cancer cells as it did in breast cancer cells. The amount of IL-32β was regulated by post-translational control rather than by transcriptional activation. Under normoxic conditions, IL-32β was continuously eliminated through ubiquitin-dependent degradation by the von-Hippel Lindau (VHL) E3 ligase complex. Oxygen deficiency or reactive oxygen species (ROS) disrupted the interaction between IL-32β and VHL, leading to the accumulation of the cytokine. The fact that IL-32β is regulated by the energy-consuming ubiquitination system implies that it plays an important role in oxidative stress. We found that IL-32β reduced protein kinase Cδ (PKCδ)-induced apoptosis under oxidative stress. This implies that the hypoxia- and ROS-stabilized IL-32β contributes to sustain survival against PKCδ-induced apoptosis.