EMAIL THIS PAGE TO A FRIEND
PMID 29050256
Abstract
As the most frequent fungal pathogen in humans,
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9384
Suloctidil, analytical standard, for drug analysis
C20H35NOS