EMAIL THIS PAGE TO A FRIEND
Oncotarget
LncSHRG promotes hepatocellular carcinoma progression by activating
PMID 29050307
Abstract
Hepatocellular carcinoma, one of the most common cancers, leads to mass mortality worldwide currently. However, the underlying mechanism of its oncogenesis remains to be elucidated. Here we identified that a long noncoding RNA,