EMAIL THIS PAGE TO A FRIEND
Journal of food science and technology
Effect of fiber blends, total solids, heat treatment, whey protein concentrate and stage of sugar incorporation on dietary fiber-fortified
PMID 29051646
Abstract
Owing to the proven beneficial role of dietary fiber (DF) on human health, feasibility of incorporating commercially available soluble and insoluble DF preparations into rice-milk-pudding (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
TDF100A Total Dietary Fiber Assay Kit, sufficient for ~100 assays