EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Development of an on-line solid phase extraction ultra-high-performance liquid chromatography technique coupled to tandem mass spectrometry for quantification of bisphenol S and bisphenol S glucuronide: Applicability to toxicokinetic investigations.
PMID 29055528
Abstract
Regulatory measures and public concerns regarding bisphenol A (BPA) have led to its replacement by structural analogues, such as Bisphenol S (BPS), in consumer products. At present, no toxicokinetic investigations have been conducted to assess the factors determining human internal exposure to BPS for subsequent risk assessment. Toxicokinetic studies require reliable analytical methods to measure the plasma concentrations of BPS and its main conjugated metabolite, BPS-glucuronide (BPS-G). An efficient on-line SPE-UPLC-MS/MS method for the simultaneous quantification of BPS and BPS-G in ovine plasma was therefore developed and validated in accordance with the European Medicines Agency guidelines for bioanalytical method validation. This method has a limit of quantification of 3ngmL
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103039
4,4′-Sulfonyldiphenol, 98%
C12H10O4S