EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Relationship between protein molecular structural makeup and metabolizable protein supply to dairy cattle from new cool-season forage corn cultivars.
PMID 29055752
Abstract
Protein solubility, ruminal degradation and intestinal digestibility are strongly related to their inherent molecular makeup. This study was designed to quantitatively evaluate protein digestion in the rumen and intestine of dairy cattle, and estimate the content of truly metabolizable protein (MP) in newly developed cool-season forage corn cultivars. The second objective was to quantify protein inherent molecular structural characteristics using advance molecular spectroscopic technique (FT/IR-ATR) and correlate it to protein metabolic characteristics. Six new cool-season corn cultivars, including 3 Pioneer (PNR) and 3 Hyland (HL), coded as PNR-7443R, PNR-P7213R, PNR-7535R, HL-SR06, HL-SR22, HL-BAXXOS-RR, were evaluated in the present study. The metabolic characteristics, MP supply to dairy cattle, and energy synchronization properties were modeled by two protein evaluation models, namely, the Dutch DVE/OEB system and the NRC-2001 model. Both models estimated significant (P<0.05) differences in contents of microbial protein (MCP) synthesis and truly absorbable rumen undegraded protein (ARUP) among the cultivars. The NRC-2001 model estimated significant (P<0.05) differences in MP content and degraded protein balance (DPB) among the cultivars. The contents MCP, ARUP and MP were higher (P<0.05) for cultivar HL-SR06, resulting in the lowest (P<0.05) DPB. However, none of the cultivars reached the optimal target hourly effective degradability ratio [25gNg/kg organic matter (OM)], demonstrating N deficiency in the rumen. There were non-significant differences among the cultivars in molecular-spectral intensities of protein. The amide I/II ratio had a significant correlation with ARUP (r=-0.469; P<0.001) and absorbable endogenous protein (AECP
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
485861
L-Alanine-2-d, ≥98 atom % D, ≥98% (CP)
C3DH6NO2