EMAIL THIS PAGE TO A FRIEND
Frontiers in pharmacology
Anti-Alzheimer's Studies on β-Sitosterol Isolated from
PMID 29056913
Abstract
The family Polygonaceae is known for its traditional use in the management of various neurological disorders including Alzheimer's disease (AD). In search of new anti-AD drugs, β-sitosterol isolated from