EMAIL THIS PAGE TO A FRIEND
Nature communications
Bottom-up precise synthesis of stable platinum dimers on graphene.
PMID 29057957
Abstract
Supported metal clusters containing only a few atoms are of great interest. Progress has been made in synthesis of metal single-atom catalysts. However, precise synthesis of metal dimers on high-surface area support remains a grand challenge. Here, we show that Pt
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
682098
Borane-ammonia complex, 97%
H6BN