EMAIL THIS PAGE TO A FRIEND
Cancer immunology, immunotherapy : CII
NY-ESO-1- and survivin-specific T-cell responses in the peripheral blood from patients with glioma.
PMID 29058035
Abstract
The prognosis for patients with glioblastoma is grim. Ex vivo expanded tumor-associated antigen (TAA)-reactive T-cells from patients with glioma may represent a viable source for anticancer-directed cellular therapies. Immunohistochemistry was used to test the survivin (n = 40 samples) and NY-ESO-1 (n = 38 samples) protein expression in tumor specimens. T-cells from peripheral blood were stimulated with TAAs (synthetic peptides) in IL-2 and IL-7, or using a combination of IL-2, IL-15 and IL-21. CD4
Related Materials