EMAIL THIS PAGE TO A FRIEND
Molecular systems biology
Lysine acetylome profiling uncovers novel histone deacetylase substrate proteins in
PMID 29061669
Abstract
Histone deacetylases have central functions in regulating stress defenses and development in plants. However, the knowledge about the deacetylase functions is largely limited to histones, although these enzymes were found in diverse subcellular compartments. In this study, we determined the proteome-wide signatures of the RPD3/HDA1 class of histone deacetylases in