EMAIL THIS PAGE TO A FRIEND
Anticancer research
Quantitative Structure-Cytotoxicity Relationship of Newly Synthesized Piperic Acid Esters.
PMID 29061797
Abstract
Eleven piperic acid esters were subjected to quantitative structure-activity relationship (QSAR) analysis based on their cytotoxicity and tumor-specificity, in order to find their new biological activities. Cytotoxicity against four human oral squamous cell carcinoma cell lines and three oral normal mesenchymal cells was determined by the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) method. Tumor specificity (TS) was evaluated by the ratio of the mean 50% cytotoxic concentration (CC One phenylmethyl ester and five phenylethyl esters showed relatively higher cytotoxicity and tumor specificity, that were significantly modified by introduction of hydroxyl and methoxy groups. On the other hand, phenylpropyl ester, phenylbutyl ester and decyl ester were essentially inactive. (2E,4E)-5-(3,4-methylenedioxyphenyl)-2,4-pentadienoic acid 2-(3,4-dihydroxyphenyl)ethyl ester [ Chemical modification of the lead compound may be a potential choice for designing a new type of anticancer drugs.