EMAIL THIS PAGE TO A FRIEND
Frontiers in cellular and infection microbiology
Activation of NF-
PMID 29062811
Abstract
In this study, we explore the regulatory roles of pro-inflammatory cytokine tumor necrosis factor alpha (TNF) in the innate immunity of macrophages against