EMAIL THIS PAGE TO A FRIEND
Frontiers in chemistry
Micropropagation and Subsequent Enrichment of Carotenoids, Fatty Acids, and Tocopherol Contents in
PMID 29062834
Abstract
A promising micropropagation protocol has been systematically established and demonstrated for the enhanced production of carotenoids, tocopherol and fatty acids in shoot tissues of
Related Materials