EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
On the nanosecond proton dynamics in phosphoric acid-benzimidazole and phosphoric acid-water mixtures.
PMID 29063940
Abstract
The unique proton conduction mechanism of phosphoric acid is important for the functions of complex phosphate containing biological and technological systems (e.g. phospholipid membranes and polybenzimidazole phosphoric acid membranes for high-temperature PEM fuel cells). In neat phosphoric acid structural proton diffusion, i.e. proton hopping between phosphoric acid molecules, is superimposed onto hydrodynamic diffusion of the molecules in the viscous liquid. In this study we separate the two dynamic contributions on the nanosecond timescale for the model systems phosphoric acid-water and phosphoric acid-benzimidazole. We demonstrate that
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194123
Benzimidazole, 98%
C7H6N2