EMAIL THIS PAGE TO A FRIEND
Scientific reports
Structural basis for the bacterial membrane insertion of dermcidin peptide, DCD-1L.
PMID 29066724
Abstract
Human dermcidin (DCD) is an antimicrobial peptide secreted constitutively by sweat glands. The anionic derivative, DCD-1L, comprises of the N-terminal 47 residues of DCD and one additional leucine residue. A previous NMR structure of DCD-1L in 50% TFE showed a partial helical conformation, and its crystal structure in the presence of Zn