EMAIL THIS PAGE TO A FRIEND
Scientific reports
Peripheral CD19
PMID 29066741
Abstract
Systemic Lupus Erythematosus (SLE) and pemphigus are two representative autoimmune diseases driven by pathogenic autoantibody systemically and organ-specifically, respectively. Given the involvement of antibody in the pathogenesis, B cells are inclined to differentiate and function in an abnormal activation model. Here we defined a unique CD19
Related Materials