EMAIL THIS PAGE TO A FRIEND
Scientific reports
Interaction Between the Trace Amine-Associated Receptor 1 and the Dopamine D
PMID 29066851
Abstract
Recent evidence suggests that the trace amine-associated receptor 1 (TAAR1) plays a pivotal role in the regulation of dopamine (DA) transmission and cocaine's actions. However, the underlying mechanisms through which TAAR1 activation mediates these effects have not yet been elucidated. Here, we used fast-scan cyclic voltammetry to measure DA dynamics and explore such mechanisms. We show, first, that the full TAAR1 agonist, RO5256390, dose-dependently blocked cocaine-induced inhibition of DA clearance in slices of the nucleus accumbens. Second, subthreshold inhibition of PKA or PKC phosphorylation did not prevent TAAR1 suppression of cocaine effects whereas subeffective doses of the DA D
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K3761
KT 5720, ≥98% (HPLC), powder
C32H31N3O5