EMAIL THIS PAGE TO A FRIEND
Frontiers in neuroscience
Optimization of
PMID 29066943
Abstract
Huntington's disease (HD) is a devastating neurodegenerative disorder caused by a polyglutamine (polyQ) expansion in exon 1 of the