EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Control of reactivity through chemical order in very small RuRe nanoparticles.
PMID 29067366
Abstract
The choice of suitable organometallic precursors, [Re
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
683353
Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)
C16H26Ru